Key Documents
BR156642
BRAND® tray stackable
size 300 x 240 x 70 mm
Synonym(s):
Photographoc trays, Trays
Select a Size
1 EA
$32.90
$32.90
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$32.90
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
$32.90
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
polypropylene
white
packaging
pack of 1 ea
manufacturer/tradename
BRAND 156642
size
300 x 240 x 70 mm
General description
The BRAND® stackable tray is made from durable white polypropylene (PP), designed for versatile use in laboratory and industrial settings. Its ribbed bottom enhances stability, preventing contents from shifting during transport or storage. The reinforced rim adds strength, while the slightly sloping walls facilitate easy access and pouring. This stackable design allows for efficient storage, maximizing space when not in use. Ideal for organizing and handling various materials, this tray combines practicality with durability, making it an essential tool for any workspace.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)