Key Documents
HS120553
Magnetic Stir Bar Retriever
white PTFE stir bar(s), L 300 mm
Synonym(s):
Magnetic Stirrer Retriever, Stir Bar Retriever
Select a Size
5 EACH
$145.00
$145.00
Estimated to ship onMay 28, 2025
Select a Size
Change View
5 EACH
$145.00
About This Item
UNSPSC Code:
41122401
NACRES:
NB.72
$145.00
Estimated to ship onMay 28, 2025
material
white PTFE stir bar(s)
packaging
pack of 5 ea
manufacturer/tradename
Heathrow Scientific 120553
L
300 mm
General description
The Stir Bar Retriever is encapsulated in PTFE making it easy to clean and durable.
Features and Benefits
  • The retrieval bar can recover stir bars even in hot solutions when stirring and mixing is no longer required
  • Chemically inert and easy to clean
  • Encapsulation in PTFE
Choose from one of the most recent versions:
Certificates of Analysis (COA)