Skip to Content
Merck
All Photos(1)

Documents

553638

Sigma-Aldrich

4-Fluorobiphenyl

95%

Synonym(s):

(4-Fluorophenyl)benzene, 1-Fluoro-4-phenylbenzene, 4-Fluorbiphenyl, 4-Fluoro-1,1′-biphenyl, 4′-Fluoro-1,1′-biphenyl, 4′-Fluorobiphenyl, p-Fluorobiphenyl, p-Fluorodiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4F
CAS Number:
Molecular Weight:
172.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

refractive index

n20/D 1.5200 (lit.)

bp

283-284 °C (lit.)

mp

75-79 °C (lit.)

density

1.288 g/mL (lit.)

SMILES string

Fc1ccc(cc1)-c2ccccc2

InChI

1S/C12H9F/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9H

InChI key

RUYZJEIKQYLEGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Fluorobiphenyl is a fluorinated biphenyl compound. It undergoes biochemical degradation in the presence of various mycorrhizal fungi to afford 4-fluorobiphen-4′-ol and 4-fluorobiphen-3′-ol as major products.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

N A Green et al.
Applied and environmental microbiology, 65(9), 4021-4027 (1999-09-03)
The pathways of biotransformation of 4-fluorobiphenyl (4FBP) by the ectomycorrhizal fungus Tylospora fibrilosa and several other mycorrhizal fungi were investigated by using (19)F nuclear magnetic resonance (NMR) spectroscopy in combination with (14)C radioisotope-detected high-performance liquid chromatography ((14)C-HPLC). Under the conditions
Cormac D Murphy
Omics : a journal of integrative biology, 11(3), 314-324 (2007-09-22)
Fluorinated organic compounds, although rare in nature, are significant environmental contaminants owing to the numerous applications for which this class of compounds is employed. It is important that biodegradation of these compounds can be readily assessed in order to provide
N A Green et al.
Chemosphere, 38(5), 1085-1101 (1999-02-24)
The incubation of the model pollutant [U-14C]'-4-fluorobiphenyl (4FBP) in soil, in the presence and absence of biphenyl (a co-substrate), was carried out in order to study the qualitative disposition and fate of the compound using 14C-HPLC and 19F NMR spectroscopy.
Jessica Amadio et al.
Applied microbiology and biotechnology, 86(1), 345-351 (2009-12-04)
The fungus Cunninghamella elegans is a useful model of human catabolism of xenobiotics. In this paper, the biotransformation of fluorinated biphenyls by C. elegans was investigated by analysis of the culture supernatants with a variety of analytical techniques. 4-Fluorobiphenyl was
Drazenka Selesi et al.
FEMS microbiology ecology, 68(1), 86-93 (2009-02-04)
The aromatic hydrocarbon biphenyl is a widely distributed environmental pollutant. Whereas the aerobic degradation of biphenyl has been extensively studied, knowledge of the anaerobic biphenyl-oxidizing bacteria and their biochemical degradation pathway is scarce. Here, we report on an enrichment culture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service