Skip to Content
Merck
All Photos(3)

Key Documents

B58601

Sigma-Aldrich

4-Bromobenzophenone

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$75.30
100 G
$86.20

$75.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$75.30
100 G
$86.20

About This Item

Linear Formula:
BrC6H4COC6H5
CAS Number:
Molecular Weight:
261.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$75.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

bp

350 °C (lit.)

mp

79-84 °C (lit.)

SMILES string

Brc1ccc(cc1)C(=O)c2ccccc2

InChI

1S/C13H9BrO/c14-12-8-6-11(7-9-12)13(15)10-4-2-1-3-5-10/h1-9H

InChI key

KEOLYBMGRQYQTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satish Kumar Talloj et al.
Journal of materials chemistry. B, 8(33), 7483-7493 (2020-07-16)
Supramolecular fluorescent materials with aggregation-induced emission (AIE) characteristics have promising applications as fluorescent probes for bio and chemosensors. In this study, a versatile low molecular weight tetraphenylethylene dipeptide hydrogelator (TPE-YY) with efficient AIE characteristics was developed, which was able to
Jun Wang et al.
Faraday discussions, 196, 335-350 (2016-11-29)
Fluorophores with aggregation-induced emission (AIE) characteristics are attractive and versatile tools for both chemical sensing and biological imaging. Herein, we designed and synthesized a fluorescent light-up system CTPE-Tau with enhanced cellular uptake ability. The system possesses several advantages, such as
Qingkai Qi et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(3), 1149-1155 (2014-11-18)
Two tetraphenylethene (TPE)-functionalized spiropyran (SP) molecules with very similar structure were designed and synthesized. The two molecules exhibit aggregation-induced emission (AIE) properties, as well as multistimuli-responsive color-changing properties, such as photochromism and acidchromism. The investigation of their different photochromic and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service