Merck
  • Reversible multistimuli-response fluorescent switch based on tetraphenylethene-spiropyran molecules.
Reversible multistimuli-response fluorescent switch based on tetraphenylethene-spiropyran molecules.
Chemistry (Weinheim an der Bergstrasse, Germany) (2014-11-18)
Qingkai Qi, Jingyu Qian, Suqian Ma, Bin Xu, Sean Xiao-An Zhang, Wenjing Tian
ABSTRACT
Two tetraphenylethene (TPE)-functionalized spiropyran (SP) molecules with very similar structure were designed and synthesized. The two molecules exhibit aggregation-induced emission (AIE) properties, as well as multistimuli-responsive color-changing properties, such as photochromism and acidchromism. The investigation of their different photochromic and acidchromic characteristics and dual-response fluorescent switch during isomerization indicated that the different link position between TPE and SP will significantly affect the extended π-conjugated system, resulting in completely different photochromic and acidchromic properties.
Product Description
Supelco
Benzophenone, analytical standard
Sigma-Aldrich
4-Bromobenzophenone, 98%
Sigma-Aldrich
Benzophenone, ReagentPlus®, 99%
Sigma-Aldrich
Benzophenone, purified by sublimation, ≥99%
Benzophenone, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzophenone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 47-49°C, analytical standard
Supelco
Mettler-Toledo Calibration substance ME 18870, Benzophenone, traceable to primary standards (LGC)
Sigma-Aldrich
1,3-Propylene sulfite, 99%