Skip to Content
Merck
All Photos(3)

Key Documents

C25007

Sigma-Aldrich

4-Chlorobenzophenone

99%

Synonym(s):

(4-Chlorophenyl)phenylmethanone

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$33.90
100 G
$77.50

$33.90


Estimated to ship onApril 04, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$33.90
100 G
$77.50

About This Item

Linear Formula:
ClC6H4COC6H5
CAS Number:
Molecular Weight:
216.66
Beilstein:
512043
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$33.90


Estimated to ship onApril 04, 2025


Request a Bulk Order

Quality Level

Assay

99%

form

crystals

bp

195-196 °C/17 mmHg (lit.)

mp

74-76 °C (lit.)

SMILES string

Clc1ccc(cc1)C(=O)c2ccccc2

InChI

1S/C13H9ClO/c14-12-8-6-11(7-9-12)13(15)10-4-2-1-3-5-10/h1-9H

InChI key

UGVRJVHOJNYEHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongkun Yu et al.
Chemistry, an Asian journal, 14(19), 3350-3356 (2019-08-27)
Eutectic molecular liquids (EMLs) based on hydrogen-bonding interaction and π-π stacking were prepared. We found that the thermodynamic properties like initial decomposition temperature and glass transition temperature of EMLs are mainly dominated by the hydrogen bond donor, which is beneficial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service