Skip to Content
Merck
All Photos(1)

Key Documents

ALD00625

Sigma-Aldrich

PSIO

Synonym(s):

(2S,3aS,6R,7aS)-3a-Methyl-2-((4-bromophenyl)thio)-6-(prop-2-yl)hexahydrobenzo[d][1,3,2]oxathiaphosphole 2-oxide

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$488.00
1 G
$681.00

$488.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$488.00
1 G
$681.00

About This Item

Empirical Formula (Hill Notation):
C16H22BrO2PS2
CAS Number:
Molecular Weight:
421.35
UNSPSC Code:
12141909
NACRES:
NA.22

$488.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

storage temp.

2-8°C

Quality Level

SMILES string

CC(C)[C@@H]1CC[C@@]2(C)[C@@H](O[P@](S2)(SC3=CC=C(Br)C=C3)=O)C1

InChI

1S/C16H22BrO2PS2/c1-11(2)12-8-9-16(3)15(10-12)19-20(18,22-16)21-14-6-4-13(17)5-7-14/h4-7,11-12,15H,8-10H2,1-3H3/t12-,15+,16+,20?/m1/s1

InChI key

FECFSGBUDPGGDZ-UMXMUBITSA-N

Application

PSIO is a P(V) reagent developed by the Baran Lab as a part of the Ψ platform, used in the direct phosphorylation of alcohols. This reagent is scalable and easy to use under mild conditions, and is functional for a broad substrate scope, including many alcohols that are challenging to otherwise activate.

Learn more about PI and PSI reagents

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

PI and PSI reagents are a platform of phosphate-based reagents for phosphorylation and phosphothiolation of nucleotides and peptides developed by the Baran lab.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service