Skip to Content
Merck
All Photos(1)

Key Documents

68572

Sigma-Aldrich

Dibutyl phosphate

≥97.0% (T)

Synonym(s):

Phosphoric acid dibutyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$63.40
1 L
$216.00

$63.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$63.40
1 L
$216.00

About This Item

Linear Formula:
(CH3CH2CH2CH2O)2P(O)OH
CAS Number:
Molecular Weight:
210.21
Beilstein:
607224
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$63.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥97.0% (T)

form

liquid

density

1.06 g/mL at 20 °C (lit.)

functional group

phosphate

SMILES string

CCCCOP(O)(=O)OCCCC

InChI

1S/C8H19O4P/c1-3-5-7-11-13(9,10)12-8-6-4-2/h3-8H2,1-2H3,(H,9,10)

InChI key

JYFHYPJRHGVZDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibutyl phosphate, also known as Phosphoric acid dibutyl ester, is an organophosphate compound with a long alkyl chain that is commonly used as an organocatalyst for polymer synthesis via ring-opening polymerization of cyclic esters and to catalyzed transesterification reactions.

Application

Dibutyl phosphate can be used as a reactant to synthesize:
  • Glycosyl phosphates by using 1,2-orthoesters.
  • 2-Aminophosphatesvia catalyst-free regioselective and enantiospecific SN2-type ring opening reaction with aziridines.
It can also be used as a catalyst to synthesize polyesters via ring-opening polymerization.

Features and Benefits

  • Inherently biodegradable
  • Stable in neutral, acidic, or alkaline solutions

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

352.4 °F - closed cup

Flash Point(C)

178 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Nishimura et al.
Journal of biochemistry, 118(1), 46-55 (1995-07-01)
Organophosphate compounds are known to cause a selective increase of beta-glucuronidase activity in rat serum. Previous data suggested that increase of serum beta-glucuronidase activity was well correlated with decrease of that activity in rat liver microsomal fraction, thereby, suggesting a
Neal B Gallagher et al.
Applied spectroscopy, 60(7), 713-722 (2006-07-21)
Multivariate curve resolution (MCR) is a powerful technique for extracting chemical information from measured spectra of complex mixtures. A modified MCR technique that utilized both measured and second-derivative spectra to account for observed sample-to-sample variability attributable to changes in soil
G K Hemakanthi De Alwis et al.
Journal of analytical toxicology, 32(9), 721-727 (2008-11-22)
Organophosphorus (OP) pesticides are highly toxic but used commonly worldwide, nevertheless. Their urinary dialkylphosphate (DAP) metabolites are widely used for exposure assessment of OP pesticides in humans. We previously developed an analytical method to measure urinary DAPs utilizing solid-phase extraction
Anticorrosive and antimicrobial efficiency of photopolymerizable phosphorus (meth) acrylate oligomers-based coating materials
Agata K, et al.
Progress in Organic Coatings, 187, 108141-108141 (2024)
Liqin Hu et al.
Chemosphere, 233, 724-732 (2019-06-15)
Organophosphate flame retardants and plasticizers (OPFRs) are widely additives in consumer products and building materials. They are frequently detected in environmental media, including indoor air, water, soil, and dust. To provide a low-cost and multi-target tool for monitoring individual exposure

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service