Skip to Content
Merck
All Photos(1)

Key Documents

924555

Sigma-Aldrich

Dimethyl phosphate

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$183.00

$183.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$183.00

About This Item

Empirical Formula (Hill Notation):
C2H7O4P
CAS Number:
Molecular Weight:
126.05
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.22

$183.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

liquid

functional group

phosphate

storage temp.

2-8°C

SMILES string

[P](=O)(OC)(OC)O

InChI

1S/C2H7O4P/c1-5-7(3,4)6-2/h1-2H3,(H,3,4)

InChI key

KKUKTXOBAWVSHC-UHFFFAOYSA-N

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service