Skip to Content
Merck
All Photos(3)

Key Documents

A7782

Sigma-Aldrich

8-Amino-2-naphthalenesulfonic acid

≥97%

Synonym(s):

1-Naphthylamine-7-sulfonic acid, Cleves acid-1,7

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$97.80
100 G
$260.00

$97.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$97.80
100 G
$260.00

About This Item

Linear Formula:
H2NC10H6SO3H
CAS Number:
Molecular Weight:
223.25
Beilstein:
2115629
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$97.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97%

form

powder

mp

≥300 °C (lit.)

SMILES string

Nc1cccc2ccc(cc12)S(O)(=O)=O

InChI

1S/C10H9NO3S/c11-10-3-1-2-7-4-5-8(6-9(7)10)15(12,13)14/h1-6H,11H2,(H,12,13,14)

InChI key

QEZZCWMQXHXAFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

8-Amino-2-naphthalenesulfonic acid can be used as a reactant to synthesize:
  • Fe3O4/sulfonated polyaniline composite material via chemical oxidative polymerization with aniline in the presence of magnetite (Fe3O4) nanoparticles.[1]
  • Prismatic crystals of strychnine-8-ammonio-2-naphthalenesulfonate-water by heating with strychnine in water/ethanol solvent mixture.[2]
  • 5-amino-9-dialkylamino benzo[a]phenoxazine dyes (Nile blue analogs) by condensation with N-alkyl or N-sulfo-propyl 4-arylazo-substituted 3-hydroxyaniline.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Strychnine-8-ammonio-2-naphthalenesulfonate-water (1/1/3.5): The first structure of a strychnine or brucine compound with a zwitterionic species
Smith G, et al.
Journal of Chemical Crystallography, 36(12), 805-811 (2006)
Synthesis and properties of magnetite/poly (aniline-co-8-amino-2-naphthalenesulfonic acid)(SPAN) nanocomposites
Reddy KR, et al.
Polymers For Advanced Technologies, 18(1), 38-43 (2007)
Development of water-soluble far-red fluorogenic dyes for enzyme sensing
Ho Nan-hui, et al.
Tetrahedron, 62(4), 578-585 (2006)
A S Rodrigues et al.
Chemosphere, 205, 433-442 (2018-05-01)
The electrodegradation of the 4-aminonaphthalene-1-sulfonic acid (4AN1S), 5-aminonaphthalene-2-sulfonic acid (5AN2S) and 8-aminonaphthalene-2-sulfonic acid (8AN2S) was studied, using two electrode materials as anode, BDD and Ti/Pt/PbO2, and two different electrolytes, sodium sulfate and sodium chloride. The highest COD removal rates were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service