Skip to Content
Merck
All Photos(2)

Key Documents

398969

Sigma-Aldrich

4-Amino-3-hydroxy-1-naphthalenesulfonic acid

ACS reagent, ≥90%

Synonym(s):

1-Amino-2-naphthol-4-sulfonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$254.00

$254.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
25 G
$254.00

About This Item

Linear Formula:
H2NC10H5(OH)SO3H
CAS Number:
Molecular Weight:
239.25
Beilstein:
2697469
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$254.00


In StockDetails


Request a Bulk Order

grade

ACS reagent

Quality Level

Assay

≥90%

form

powder

ign. residue

≤0.1%

mp

290 °C (dec.) (lit.)

anion traces

sulfate (SO42-): ≤0.2%

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Nc1c(O)cc(c2ccccc12)S(O)(=O)=O

InChI

1S/C10H9NO4S/c11-10-7-4-2-1-3-6(7)9(5-8(10)12)16(13,14)15/h1-5,12H,11H2,(H,13,14,15)

InChI key

RXCMFQDTWCCLBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Amino-3-hydroxy-1-naphthalenesulfonic acid is a derivative of aniline consisting of a trifunctional monomer (bearing three functional groups –NH2, –OH and –SO3H). It also contains two fused benzene rings.

Application

4-Amino-3-hydroxy-1-naphthalenesulfonic acid has been used as a probe in a novel fluorescence assay for the determination of parathion-methyl.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service