Skip to Content
Merck
All Photos(1)

Key Documents

930024

Sigma-Aldrich

Hyaluronic acid adipic dihydrazide (HA-ADH)

Synonym(s):

HA-ADH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C20H33N5O12)n
UNSPSC Code:
12352125
NACRES:
NA.23

form

solid (Chunks)

Quality Level

color

white to off-white

application(s)

3D bioprinting

storage temp.

2-8°C

General description

Adipic acid dihydrazide is commonly used as a functional group to allow for HA cross-linking to polymers containing aldehyde groups or that are aldehyde-functionalized, like HA-aldehyde or PEG-aldehyde. These biomaterials are attractive for their tunable properties and low cytotoxicity.
Degree of functionalization (50-80%)

Application

Hyaluronic acid (HA) is a ubiquitous extracellular matrix component, which makes it attractive for use in medical applications for it′s biocompatibility and low immunogenicity. HA-based polymers are used to make hydrogels for a wide variety of applications from tissue engineering and regeneration to drug delivery and bioinks for 3D bioprinting. To make hydrogels, HA has to be derivatized with groups that can cross-link to to other functionalized polymers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuguo Lei et al.
Biomaterials, 32(1), 39-47 (2010-10-12)
Synthetic hydrogel scaffolds that can be used as culture systems that mimic the natural stem cell niche are of increased importance for stem cell biology and regenerative medicine. These artificial niches can be utilized to control the stem cell fate
Xinqiao Jia et al.
Biomacromolecules, 7(12), 3336-3344 (2006-12-13)
Vocal fold scarring disrupts the viscoelastic properties of the lamina propria that are critical for normal phonation. There is a clinical need for the development of advanced biomaterials that approximate the mechanical properties of the lamina propria for in vivo
Self-crosslinking and injectable hyaluronic acid/RGD-functionalized pectin hydrogel for cartilage tissue engineering
Chen, et al.
Carbohydrate Polymers, 166, 31-44 (2017)

Articles

Engineered ECMs enhance immune therapy in cancer treatment by supporting cells and tissues and modulating immune response. They improve immune cell maturation, expansion, and regulation through biomaterial manipulation, acting as frameworks or carriers for enhanced tumor immunotherapy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service