Skip to Content
Merck
All Photos(1)

Key Documents

927279

Sigma-Aldrich

Hyaluronic acid tetrabutylammonium (HA-TBA)

mol wt 30 kDa

Synonym(s):

HA-TBA, Hyaluronan tetrabutylammonium, Hyaluronic acid tetrabutylammonium, Tetrabutylammonium -Modified Hyaluronan, Tetrabutylammonium functionalized hyaluronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
$236.00

$236.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 G
$236.00

About This Item

Linear Formula:
[C30H56N2O11]n
UNSPSC Code:
12352106
NACRES:
NA.25
form:
(solid or chunk(s) or fiber )

$236.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

(solid or chunk(s) or fiber )

Quality Level

mol wt

30 kDa

color

white to off-white

storage temp.

2-8°C

Application

HA is a naturally occurring unbranched glycosaminoglycan (GAG), a polyanionic polymer of high molecular weight that exhibits unique viscoelastic properties, excellent biocompatibility, and biodegradability. These properties of HA-derived hydrogels make them ideal biomaterials for tissue engineering. Hyaluronic acid tetrabutylammonium is a simple and effective way to make hyaluronic acid soluble in organic solvents, which is a necessary step for a wide variety of functionalizations.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julianne L Holloway et al.
Journal of controlled release : official journal of the Controlled Release Society, 191, 63-70 (2014-06-07)
Bone morphogenetic proteins (BMPs) show promise in therapies for improving bone formation after injury; however, the high supraphysiological concentrations required for desired osteoinductive effects, off-target concerns, costs, and patient variability have limited the use of BMP-based therapeutics. To better understand

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service