Skip to Content
Merck
All Photos(1)

Key Documents

924474

Sigma-Aldrich

Hyaluronic Acid

Low Viscosity, Low endotoxin

Synonym(s):

3D bioprinting, HA, Hyaluronan

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$270.00

$270.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$270.00

About This Item

UNSPSC Code:
12352201
NACRES:
NA.23

$270.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

(powder or chunk(s) or fibers)

impurities

<10 CFU/g Bioburden
<100 EU/g Endotoxin

color

white to off-white

suitability

conforms to structure for NMR

storage temp.

2-8°C

General description

Hyaluronic acid (HA) is a linear polysaccharide of alternating D-glucuronic acid, and N-acetyl-D-glucosamine found in the extracellular matrix. HA is commonly chemically modified to form covalently crosslinked hydrogels. Other properties of HA include cell surface receptors CD44 and RHAMM recognition and degradation via hyaluronidases.

Application

HA is increasingly used for the following biomedical applications:
  • Cell migration
  • Angiogenesis
  • Viability
  • Proliferation

Our low endotoxin HA is tested for both endotoxin (< 100 EU/g) and bioburden (< 10 CFU/g) levels, so you can choose which material is right for your research.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federica Camin et al.
Journal of agricultural and food chemistry, 58(1), 570-577 (2009-12-17)
H, C, and O stable isotope ratios and the elemental profile of 267 olive oils and 314 surface waters collected from 8 European sites are presented and discussed. The aim of the study was to investigate if olive oils produced
Xian Xu et al.
Soft matter, 8(12), 3280-3294 (2012-03-16)
Hyaluronic acid (HA) is one of nature's most versatile and fascinating macromolecules. Being an essential component of the natural extracellular matrix (ECM), HA plays an important role in a variety of biological processes. Inherently biocompatible, biodegradable and non-immunogenic, HA is
Tracey J Brown
Current pharmaceutical biotechnology, 9(4), 253-260 (2008-08-12)
Despite advances in chemotherapeutic regimens, the treatment of metastatic cancer remains a challenge. A key problem with chemotherapy drugs is nonspecific drug distribution, resulting in low tumor concentrations and systemic toxicity. The holy grail of clinical cancer research has been
Kevin T Dicker et al.
Acta biomaterialia, 10(4), 1558-1570 (2013-12-24)
Hyaluronan (HA) is a linear polysaccharide with disaccharide repeats of d-glucuronic acid and N-acetyl-d-glucosamine. It is evolutionarily conserved and abundantly expressed in the extracellular matrix (ECM), on the cell surface and even inside cells. Being a simple polysaccharide, HA exhibits
Jason A Burdick et al.
Advanced materials (Deerfield Beach, Fla.), 23(12), H41-H56 (2011-03-12)
Hyaluronic acid (HA), an immunoneutral polysaccharide that is ubiquitous in the human body, is crucial for many cellular and tissue functions and has been in clinical use for over thirty years. When chemically modified, HA can be transformed into many

Articles

Engineered ECMs enhance immune therapy in cancer treatment by supporting cells and tissues and modulating immune response. They improve immune cell maturation, expansion, and regulation through biomaterial manipulation, acting as frameworks or carriers for enhanced tumor immunotherapy.

Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. what MW range is the HA in this poduct?

    1 answer
    1. The molecular weight of this product is not determined. However, historical data suggests batches typically have a Mn (molecular number) around 20-40kDa.

      Helpful?

  4. How can this product best be reconstituted for surface coating TC substrates?

    1 answer
    1. Unfortunately, this product is not tested for solubility. However, according to the literature, Hyaluronic acid is soluble in water. Dissolve in cold water and shake slowly until the hyaluronic acid is dissolved completely. Please note this has not been validated internally.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service