Skip to Content
Merck
All Photos(1)

Key Documents

923575

Sigma-Aldrich

Silver nano ink

Screen printable, Photonically curable

Synonym(s):

Ag NP-based ink, Photonically curable Ag ink, S-CS21306, Screen printable Ag ink, Silver (Ag) Nanoparticle Ink, Silver nano ink

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$412.00

$412.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$412.00

About This Item

Linear Formula:
Ag
UNSPSC Code:
12352103
NACRES:
NA.23

$412.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

sheet resistance

15.6m Ω/sq, 25 mum after curing at 150 Celsius for 5 min on glass
7.5m Ω/sq, 25 mum after curing at 200 Celsius for 5 min on glass
75m Ω/sq, 25 mum after curing at 120 Celsius for 1h on glass

viscosity

6500-10000 mPa.s(20 °C) (@100/s)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Screen printing silver nano ink can be utilized as a precursor to produce conductive layers for electroluminescent displays. These displays are used in applications such as lighting panels, automotive displays, and signage. It can be used in studies such as flexible circuitry, sensors, touch panels, RFID antennas, and other electronic components.
This Smart’Ink S-CS21306 conductive silver ink is particularly well suited for applications requiring high conductivity and high resolution at very low curing temperatures. This ink, based on silver, is perfectly adapted to design conductive traces on flexible substrates. It is suitable for IoT applications such as antennas (HF, UHF).
Process: Rotary screen printing (below 80μm of resolution)

This ink has been developed to allow photonical curing to enable high throughput printing.

Compatible Printing Method: screen printing,
Compatible substrates: PET (Melinex 406, Melinex 339, Melines 520, Arcophane TCA, Arcophane STS, Folex BG-71), PEN (Teonex), Kapton® , Ceramic
Recommended surface treatment for substrates:
  • Temperature stabilization
  • Argon plasma treatment

Features and Benefits

  • Good bending resistance, smooth surface
  • Superior conductivity
  • High resolution
  • Curing process compatibility: photonic, NIR, low vacuum oven, thermal curing
  • High nano-particle content
  • Non-toxic (No CMR ink)

Preparation Note

Printing guidelines:

  • Inox, 18 µm mesh thickness, 640/inch-250/cm mesh count, 45 degree mesh angle, 7 µm emulsion thickness, 50 µm resolution, 83 m2/kg theoretical coverage, 1 µm theoretical dried thickness.
  • Inox, 19 µm mesh thickness, 500/inch-200/cm mesh count, 22.5 degree mesh angle, 5 µm emulsion thickness, 40 µm resolution, 80 m2/kg theoretical coverage, 1 µm theoretical dried thickness.
  • Polyester, 30 µm mesh thickness, 380/inch-150/cm mesh count, 22.5 degree mesh angle, 10 µm emulsion thickness, 80 µm resolution, 50 m2/kg theoretical coverage, 1.6 µm theoretical dried thickness.

Legal Information

Kapton is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service