Skip to Content
Merck
All Photos(2)

Key Documents

796042

Sigma-Aldrich

Silver nanoparticle ink

50 wt. %, dispersion in tripropylene glycol mono methyl ether

Synonym(s):

Ag ink, Conductive ink, Silver ink

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$262.00
20 G
$706.00

$262.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$262.00
20 G
$706.00

About This Item

Linear Formula:
Ag
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$262.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

concentration

50 wt. %

resistivity

≤20 μΩ-cm (after sintering at 200°C; 2h)
≤7 μΩ-cm (after sintering at 240°C; 4h)

particle size

115 nm (d90)
70 nm (d50)

surface tension

24 dyn/cm

viscosity

24 cP

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver is a good conductor and resistant to oxidation. It is used in printed conductors and printable inks (based on silver nanoparticles). Nanosilver is used as an antibacterial material in filters for air and water purification, in textiles, and for wound treatment. Other large scale applications of nanosilver include metallization of solar cells, thin film electronics, sensors, and catalysts.[1]

Application

  • The ink is suitable for digital printing such as Inkjet and Aerosol systems.
  • Photovoltaic Cells
  • Printed Electronics: FPD; RFID; PCB
  • Decoration

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nanostructured Materials Through Ultrasonic Spray Pyrolysis
Bang JH, et al.
Material Matters, 7(2) null

Articles

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

Progress in solution-processed functional materials leads to thin-film optoelectronic devices for industrial and consumer electronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service