Skip to Content
Merck
All Photos(1)

Key Documents

735825

Sigma-Aldrich

Silver

greener alternative

conductive paste

Synonym(s):

Silver Paste DGP80 TESM8020

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$409.00

$409.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$409.00

About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
11101802
PubChem Substance ID:
NACRES:
NA.23

$409.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

paste (microparticles)

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

≥75%

resistivity

1-3 * 10^-5 Ω-cm (conductive paste)
1.59 μΩ-cm, 20°C

matrix

α-Terpineol

particle size

200 nm (80%)
<5 nm (20%)

viscosity

100,000-300,000 cP, 0.4 rpm(25 °C, Brookfield)

bp

2212 °C (lit.)

mp

960 °C (lit.)

transition temp

curing temperature 120-150 °C (30-60 min)

density

10.49 g/cm3 (lit.)

greener alternative category

storage temp.

2-8°C

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

General description

Dispersion

Recommended washing solvent = polar solvent (Ethanol, IPA, Methanol, etc)

Recommended substrates = ITO film & glass, plastic film
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". 3D printable inks help to design and print more complex designs with minimum waste and it is environmentally friendly. Click here for more information.

Application

Silver conductive paste (Ag) can be used as an adhesive which can be used on indium tin oxide (ITO) for a variety of electronic applications.[1][2] It can also be used as an interdigitated electrode (IDE) for gas sensors.[3][4]
This product can be used in material extrusion 3D printing technique.

Legal Information

Product of ANP, Inc.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

205.0 °F

Flash Point(C)

96.1 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Hydrogen Gas Sensor Based on TiO2 Nanoparticles on Alumina Substrate
Mohd Chachuli S, et al.
Sensors, 18(8), 2483-2483 (2018)
Structural transformations of mechanically alloyed polycrystalline YMnO 3-based material for gas sensing application
Abdullah NH, et al.
Journal of the Australasian Ceramic Society, 18(8), 1-12 (2019)
Optimization of physical properties of vacuum evaporated CdTe thin films with the application of thermal treatment for solar cells
Chander S and Dhaka MS
Materials Science in Semiconductor Processing, 40, 708-712 (2015)
Physical properties of vacuum evaporated CdTe thin films with post-deposition thermal annealing
Chander S and Dhaka MS
Physica E: Low-Dimensional Systems and Nanostructures, 73, 35-39 (2015)
R Nirmala et al.
Journal of nanoscience and nanotechnology, 13(7), 4686-4693 (2013-08-02)
We report on the preparation and characterization of polyurethane (PU) nanofibers containing silver (Ag) nanoparticles were synthesized by using electrospinning. Two different approaches were adopted to incorporate the Ag nanoparticles in to PU nanofibers. In the first approach, a homogeneous

Articles

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

While dye sensitization as the basis for color photography has been accepted for a very long time,1 attempts to use this principle for the conversion of solar light to electricity generally had resulted only in very low photocurrents, below 100 nA/cm

Ultrasonic spray pyrolysis produces scalable nanomaterials like metal oxides and quantum dots for diverse applications.

See All

Questions

1–6 of 6 Questions  
  1. Do you have any suggestion for dilution? I want to control the viscosity. I confirmed that the dispersion matrix is alpha-terpineol. Can I use that solvent to dilute it? If so, Could you recommend the specific method? (i.e. sonication)

    1 answer
    1. Alpha-terpineol can be used for dilution, as it is a component of this product. However, no specific method or protocol for dilution is available.

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  4. Does this paste contain any silicones or siloxanes?

    1 answer
    1. This product is ≥75% Silver. The remaining content is the α-Terpineol dispersion matrix. There are no silicone or siloxane components present.

      Helpful?

  5. what is the curing temperature of silver conducting paste ?

    1 answer
    1. As stated in the 'PROPERTIES' section of the product page, the curing temperature is 120-150 °C for 30-60 min.

      Helpful?

  6. What is the duration for bonding two structures together when using this product? Additionally, how long does it take to solidify? Also, I need information on the duration until it is no longer conductive.

    1 answer
    1. The duration for solidification and conductivity can vary based on factors such as substrate type and curing temperature. Additionally, we do not have any experience or test results regarding your technical inquiry

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service