Skip to Content
Merck
All Photos(6)

Key Documents

922218

Sigma-Aldrich

Male luer plug

single, Fluidic 263, PP

Synonym(s):

Microfluidic chip accessories

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$75.90

$75.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$75.90

About This Item

UNSPSC Code:
42142600
NACRES:
NA.25

$75.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Application

The male Luer plugs enable closing of female luer and luer lock interfaces on our fluidic platforms. The plugs allow for the movement of liquid with the female luer interface into the fluidic channels on the chip, while keeping the fluidic interface closed to avoid contamination.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent advances of controlled drug delivery using microfluidic platforms.
Li X, et al.
Advanced Drug Delivery Reviews, 18, 3-28 (2018)
Microfluidic-assisted fabrication of carriers for controlled drug delivery.
Santos H A, et al.
Lab on a chip, 17, 1856-1883 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service