Skip to Content
Merck
All Photos(1)

Documents

58700-U

PTFE Tubing

L × O.D. × I.D. 10 ft × 1/16 in. × 0.031 in.

Synonym(s):

PTFE Laboratory Tubing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

packaging

pkg of 1 ea

L × O.D. × I.D.

10 ft × 1/16 in. × 0.031 in.

Looking for similar products? Visit Product Comparison Guide

General description

Economical, flexible PTFE tubing is ideal for use at pressures up to 500 psi (35 kg/cm2). Use in automatic analyzer, postcolumn reaction, preparative scale systems, in-stream sampling devices, and when monitoring physiologically important compounds.

The maximum recommended operating temperature for PTFE is 200 °C, but short term exposure to higher temperatures seldom causes damage.

Application

  • Microfluidic device for multiple emulsions: Discusses the development of a microfluidic device using PTFE tubing to generate multiple emulsions, with applications in pharmaceuticals and material science (Khalilnezhad A, Sahraei E, Cortes FB, Riazi M, 2023).
  • Cell Microencapsulation within Gelatin-PEG Microgels: Uses PTFE tubing in a simple pipet tip-based device for cell microencapsulation, important for biomedical applications such as tissue engineering (Nguyen TPT, Li F, Hung B, Truong VX, Thissen H, Forsythe JS, Frith JE, 2023).
  • Finned Tubular Air Gap Membrane Distillation: Introduces a finned tubular design with PTFE membranes for optimizing distillation processes, potentially impacting desalination and wastewater treatment (Wu Z, Guo F, 2023).
  • Expanded PTFE/Graphite Composites for Water/Oil Separation: Develops PTFE/graphite composites to enhance the efficiency of water/oil separation, offering benefits for environmental protection and industrial processes (Aturaliya R, Wang D, Xu Y, Lin YJ, Li Q, Turng LS, 2020).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service