Skip to Content
Merck
All Photos(1)

Key Documents

920444

Sigma-Aldrich

mPEG functionalized gelatin

50% PEGylation, PEG average Mn 1k, gel strength 300 g Bloom

Synonym(s):

PEGylated Gelatin

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$297.00

$297.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$297.00

About This Item

Linear Formula:
(C32H50N9O12-)m(CH2CH2O)n
UNSPSC Code:
12352201
NACRES:
NA.23

$297.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

(solid or chunk(s) or fiber)

mol wt

PEG average Mn 1k

color

white to off-white

storage temp.

2-8°C

Application

mPEG functionalized gelatin is Type A gelatin grafted with PEG that can be used in drug delivery research. Gelatin is a denatured protein that is obtained by hydrolysis of animal collagen. Its biodegradability, biocompatibility, chemical modification potential and cross-linking possibility make gelatin-based nanoparticles (GNPs) a promising carrier system for drug delivery applications. mPEG functionalization has several advantages − prolonged circulation in the body, decreased degradation by metabolic enzymes, and reduction or elimination of protein immunogencity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modified gelatin nanoparticles for gene delivery.
Wettig S D, et al.
Int. J. Pharmacol., 554, 224-234 (2019)
A novel smart PEGylated gelatin nanoparticle for co-delivery of doxorubicin and betanin: A strategy for enhancing the therapeutic efficacy of chemotherapy.
Ghorbani M, et al.
Materials Science and Engineering, C, 97, 833-841 (2019)
Ahmed O Elzoghby
Journal of controlled release : official journal of the Controlled Release Society, 172(3), 1075-1091 (2013-10-08)
Gelatin is one of the most versatile natural biopolymers widely used in pharmaceutical industries due to its biocompatibility, biodegradability, low cost and numerous available active groups for attaching targeting molecules. These advantages led to its application in the synthesis of
Sajed Amjadi et al.
Materials science & engineering. C, Materials for biological applications, 97, 833-841 (2019-01-27)
Betanin (BET) can reduce the side effects of potent anticancer drugs e.g. doxorubicin (DOX) on the normal tissues in co-administration with them because of the synergistic therapeutic effect and consequently the reduced required amount of anticancer agents. Despite interest in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service