Skip to Content
Merck
All Photos(1)

Key Documents

914509

Sigma-Aldrich

Polycaprolactone diacrylate

average Mn 10,000

Synonym(s):

PCL diacrylate, PCL-diacrylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$285.00

$285.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$285.00

About This Item

Linear Formula:
(C6H10O2)nC10H14O5
UNSPSC Code:
12162002
NACRES:
NA.23

$285.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

mol wt

average Mn 10,000

color

off-white to beige

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Polycaprolactone is a biodegradable polymer commonly used in biomedical applications. It is a semicrystalline polymer that possesses relatively long degradation time. Polycaprolactone diacrylate carriers photoreactive end groups for crosslinking, which allows its application in 3D printing and controlled drug delivery research.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maneesh Jaiswal et al.
Biomedical materials (Bristol, England), 5(6), 065014-065014 (2010-11-17)
A biodegradable semi-interpenetrating hydrogel network (semi-IPN) of polyacrylamide and gelatin was prepared using polycaprolactone diacrylate (mol. wt ∼ 640) as a crosslinker. The drug-polymer interaction and IPN formation were investigated by attenuated total reflectance-Fourier transform infrared (ATR-FTIR) and thermal gravimetric
Yinfeng He et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 105(6), 1645-1657 (2016-05-15)
A new type of photocrosslinkable polycaprolactone (PCL) based ink that is suitable for three-dimensional (3D) inkjet printing has been developed. Photocrosslinkable Polycaprolactone dimethylacrylate (PCLDMA) was synthesized and mixed with poly(ethylene glycol) diacrylate (PEGDA) to prepare an ink with a suitable

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service