Skip to Content
Merck
All Photos(1)

Key Documents

900625

Sigma-Aldrich

Polycaprolactone diol

average Mn 10,000

Synonym(s):

Diethylene glycol-initiated polycaprolactone, Polycaprolactone oxydiethylene ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$216.00

$216.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$216.00

About This Item

Linear Formula:
C4H8O3(C6H10O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$216.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

mol wt

average Mn 10,000

transition temp

Tm 57-62 °C (experimental)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Polycaprolactone diol is a biodegradable, ε-caprolactone-based polymer for use in biomedical research applications. Due to the synthetic method used, this material features two terminal hydroxyl units, allowing it to be readily functionalized or used in the synthesis of more complex polymers (e.g. polyurethanes) for applications such as shape-memory materials. Due to the high aliphatic content and molecular weight, polycaprolactone diol features increased stability in vivo and high Tg, respectively.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Injectable polymeric biomaterials: a brief review.
Dou J, et al.
Materials Technology: Advanced Performance Materials, B273-B282 (2015)
A novel degradable polycaprolactone networks for tissue engineering.
Kweon HY, et al.
Biomaterials, 801-808 (2003)
Preparation and characterization of highly porous biodegradable polyurethane scaffolds for soft tissue applications.
Guan J, et al.
Biomaterials, 3961-3971 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service