Skip to Content
Merck
All Photos(3)

Key Documents

189405

Sigma-Aldrich

Polycaprolactone diol

average Mn ~530

Synonym(s):

Diethylene glycol-initiated polycaprolactone, Polycaprolactone oxydiethylene ester

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$414.00
500 G
$763.00

$414.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$414.00
500 G
$763.00

About This Item

Linear Formula:
C4H8O3(C6H10O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$414.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

paste

Quality Level

mol wt

average Mn ~530

transition temp

softening point 35 °C

density

1.073 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Application

Polycaprolactone diol(PCL-diol) may be used as a precursor to biodegradable and bioresorbable polycaprolactone.[1] PCL-diol in combination with polytetrahydrofuran diol (PTHF-diol) and ceric ammonium nitrate may be used to initiate polymerization of methyl methacrylate (MMA).[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deposition of Biodegradable Polycaprolactone Coatings Using an In?line Atmospheric Pressure Plasma System.
Tynan J, et al.
Plasma Processes and Polymers, 6(S1), S51-S56 (2009)
Ceric ion initiation of methyl methacrylate using polytetrahydrofuran diol and polycaprolactone diol.
Arslan H and Hazer B
European Polymer Journal, 35(8), 1451-1455 (1999)

Articles

In the past two decades, tissue engineering and regenerative medicine have become important interdisciplinary fields that span biology, chemistry, engineering, and medicine.

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service