Skip to Content
Merck
All Photos(1)

Key Documents

914223

Sigma-Aldrich

Biotin-SLF

Synonym(s):

(R)-3-(3,4-Dimethoxyphenyl)-1-(3-(4,18-dioxo-22-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-8,11,14-trioxa-5,17-diazadocosanamido)phenyl)propyl (S)-1-(3,3-dimethyl-2-oxopentanoyl)piperidine-2-carboxylate, Bio-SLF, Biotin-conjugated FKBP12WT-directed ligand, Biotin-conjugated SLF ligand, FKBP12WT affinity probe for PROTAC® and targeted protein degradation research

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$877.00

$877.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$877.00

About This Item

Empirical Formula (Hill Notation):
C52H76N6O13S
CAS Number:
Molecular Weight:
1025.26
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

$877.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

ligand

biotin

Quality Level

form

powder

storage temp.

−20°C

SMILES string

O=C(CCC(NCCOCCOCCOCCNC(CCCC[C@@H]1SC[C@@](N2)([H])[C@]1([H])NC2=O)=O)=O)NC3=CC([C@H](OC([C@@H]4CCCCN4C(C(C(C)(C)CC)=O)=O)=O)CCC5=CC=C(OC)C(OC)=C5)=CC=C3

InChI key

QFIMAWUXIBINCP-SLBVAARVSA-N

Application

Biotin-SLF (bio-SLF) is an FKBP12WT affinity probe useful in chemical biology or targeted protein degradation where FKBP12 is used as a tool protein and its enrichment is required in discovery experiments or assay validation.


Related tools:
Additional bifunctional tools for FKPB12 variants:

  • dTAG-13 (SML2601) for FKBP12F36V; and KB02-SLF (914738), KB03-SLF (914975), and KB04-SLF (913715) for FKBP12WT
  • Cereblon (CRBN) affinity probe: Biotin-Thalidomide (913979)
Automate your Biotin tagging with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Behnam Nabet et al.
Nature chemical biology, 14(5), 431-441 (2018-03-28)
Dissection of complex biological systems requires target-specific control of the function or abundance of proteins. Genetic perturbations are limited by off-target effects, multicomponent complexity, and irreversibility. Most limiting is the requisite delay between modulation to experimental measurement. To enable the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service