Skip to Content
Merck
All Photos(1)

Key Documents

913480

Sigma-Aldrich

Titanium dioxide nanofibers

Synonym(s):

Nanofibrous inorganic powder, TiO2 anatase

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
TiO2
CAS Number:
Molecular Weight:
79.87
UNSPSC Code:
11181502
NACRES:
NA.23

description

Thermal Conductivity 6.5 Wm-1K-1

Quality Level

form

nanofiber
powder (3D cotton/white fluffy)

fiber diam. × L

200-800 nm ± 100 nm × 2-hundreds of μm , Structure: polycystalline nanofibers

surface area

10 m2/g

refractive index

n/D 2.5

primary crystallite size

(anatase
Typical Size of Crystallites: 7 nm)

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Application

Titania oxdie-anatase nanofibers has energy applications as energy convertor in solar cells, air/fuel ratio controllor in automotive, humidity and gas sensors, electrode materials in batteries, solid oxide fuel cells, inorganic membranes, dye sensitized solar cells, photocatalyic degradation of votile compounds (VOC), bacteria and NOx, and waste water purification.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Review discusses recent progress and challenges in ORR catalyst development for PEMFC applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service