Skip to Content
Merck
All Photos(1)

Key Documents

774510

Sigma-Aldrich

Titanium(IV) oxide

nanowires, diam. × L ~100 nm × 10 μm

Synonym(s):

Titania nanofibers, Titania nanowires, Titanium dioxide nanofibers, Titanium dioxide nanowires, Titanium(IV) oxide nanofibers, Titania, Titanium dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$466.00

$466.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$466.00

About This Item

Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

$466.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

nanowires

diam. × L

~100 nm × 10 μm

mp

>350 °C (lit.)

density

4.26 g/mL at 25 °C (lit.)

SMILES string

O=[Ti]=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Titanium dioxide nanowires have been studied as photocatalysts [1]; battery anode materials[2]; catalyst supports[3] and as a replacement for TiO2 nanospheres in dye sensitized solar cells[4].

Preparation Note

Product prepared by hydrothermal treatment.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuan Pan et al.
ACS applied materials & interfaces, 4(8), 3944-3950 (2012-07-12)
We demonstrate that uniform dispersion of TiO(2) on graphene is critical for the photocatalytic effect of the composite. The hydrothermal method was employed to synthesize TiO(2) nanowires (NW) and then fabricate graphene-TiO(2) nanowire nanocomposite (GNW). Graphene oxide (GO) reduction to
Yiping; T.;
Electrochimica Acta, 78, 154-159 (2012)
Matt Law et al.
Nature materials, 4(6), 455-459 (2005-05-17)
Excitonic solar cells-including organic, hybrid organic-inorganic and dye-sensitized cells (DSCs)-are promising devices for inexpensive, large-scale solar energy conversion. The DSC is currently the most efficient and stable excitonic photocell. Central to this device is a thick nanoparticle film that provides
Eric Formo et al.
Nano letters, 8(2), 668-672 (2008-01-22)
This paper reports a simple procedure for derivatizing the surface of anatase TiO2 nanofibers with Pt nanoparticles and then Pt nanowires. The nanofibers were prepared in the form of a nonwoven mat by electrospinning with a solution containing both poly(vinyl
Michael Bushell et al.
Nanomaterials (Basel, Switzerland), 10(9) (2020-09-17)
Physical chemical characterization of nanomaterials is critical to assessing quality control during production, evaluating the impact of material properties on human health and the environment, and developing regulatory frameworks for their use. We have investigated a set of 29 nanomaterials

Articles

Dye-sensitized solar cells (DSCs) are 3rd generation solar cells combining the promise of high efficiency with low production costs.

Among various ceramics, one-dimensional (1-D) piezoelectric ceramics have attracted significant scientific attention for use in energy harvesting.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

Catalytic water splitting produces hydrogen crucial for renewable energy, petroleum refining, and chemical industry applications like methanol production.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service