Skip to Content
Merck
All Photos(1)

Key Documents

914401

Sigma-Aldrich

Titanium dioxide milled nanofibers

anatase/rutile

Synonym(s):

Nanofibrous inorganic powder, TiO2 anatase/rutile

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$226.00
10 G
$395.00

$226.00

PRICE WITHOUT NATIONAL TAXES

0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$226.00
10 G
$395.00

About This Item

Empirical Formula (Hill Notation):
TiO2
CAS Number:
Molecular Weight:
79.87
MDL number:
UNSPSC Code:
12171604
NACRES:
NA.23

$226.00

PRICE WITHOUT NATIONAL TAXES

0 left in stock

Request a Bulk Order

description

Thermal Conductivity 6.5 Wm-1K-1

Quality Level

form

nanofiber
powder (fluffy)

fiber diam. × L

200-800 nm ± 100 nm × 2-10 μm , Structure: Polycystalline nanofibers

surface area

10 m2/g

color

white

refractive index

n/D 2.7

primary crystallite size

(anatase-rutil
Typical Size of Crystallites: 10 nm)

SMILES string

[Ti](=O)=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Application

Titania oxdie-anatase/rutil nanofibers has energy applications as energy convertor in solar cells, air/fuel ratio controllor in automotive, humidity and gas sensors, electrobe materials in batteries, solid oxide fuel cells, inorganic membranes, dye sensitized solar cells, photocatalyic degradation of votile compounds (VOC), bacteria and NOx, and waste water purification.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Review discusses recent progress and challenges in ORR catalyst development for PEMFC applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service