Skip to Content
Merck
All Photos(1)

Key Documents

767743

Sigma-Aldrich

2,5-Dihydro-3,6-di-2-thienyl-pyrrolo[3,4-c]pyrrole-1,4-dione

97%

Synonym(s):

3,6-Di(thiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione, DPP

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$1,870.00

$1,870.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$1,870.00

About This Item

Empirical Formula (Hill Notation):
C14H8N2O2S2
CAS Number:
Molecular Weight:
300.36
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$1,870.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

SMILES string

O=C1NC(c2cccs2)=C3C(=O)NC(c4cccs4)=C13

InChI

1S/C14H8N2O2S2/c17-13-9-10(12(16-13)8-4-2-6-20-8)14(18)15-11(9)7-3-1-5-19-7/h1-6H,(H,15,18)(H,16,17)

InChI key

YIUHGBNJJRTMIE-UHFFFAOYSA-N

General description

2,5-Dihydro-3,6-di-2-thienyl-pyrrolo[3,4-c]pyrrole-1,4-dione is a diketopyrrolopyrrole (DPP) based copolymer and are used widely in thin-film transistors and solar cell devices. This aromatic polymer has electron withdrawing groups and hence is very useful for the synthesis of narrow band gap donor-acceptor polymers which are used as active semiconductors for organic electronics. They give rise to high power conversion efficiency (PCE) in Organic Photovoltaic (OPV) Cells. [1]

Application

Used in the synthesis of donor-acceptor polymers which are used in polymer field-effect transistors and bulk heterojunction solar cells. [2][3]

Features and Benefits

Diketopyrrolopyrrole is planar and can accept hydrogen bonds/other electrostatic interactions which results in copolymers that have pi-pi stacking.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of phenanthrocarbazole-diketopyrrolopyrrole copolymer for high-performance field-effect transistors
Chen, H., Guo, Y., Sun, X., Gao, D., Liu, Y., & Yu, G.
Journal of Polymer Science Part A: Polymer Chemistry, 51(10), 2208-2215 (2013)
David Rais et al.
ChemPlusChem, 85(12), 2689-2703 (2020-12-18)
The singlet fission (SF) process discovered in bis(thienyl)diketopyrrolopyrroles (TDPPs) can boost their potential for photovoltaics (PV). The crystal structures of TDPP analogs carrying n-hexyl, n-butyl, or 2-(adamant-1-yl)ethyl substituents are similar, but contain increasingly slipped stacked neighbor molecules. The observed SF
Influence of Side-Chain on Structural Order and Photophysical Properties in Thiophene Based Diketopyrrolopyrroles: A Systematic Study.
Naik, M. A., Venkatramaiah, N., Kanimozhi, C., & Patil, S.
The Journal of Physical Chemistry C, 116(50), 26128-26137 (2012)
Hugo Bronstein et al.
Journal of the American Chemical Society, 133(10), 3272-3275 (2011-02-22)
We report the synthesis and polymerization of a novel thieno[3,2-b]thiophene-diketopyrrolopyrrole-based monomer. Copolymerization with thiophene afforded a polymer with a maximum hole mobility of 1.95 cm(2) V(-1) s(-1), which is the highest mobility from a polymer-based OFET reported to date. Bulk-heterojunction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service