Skip to Content
Merck
All Photos(2)

Key Documents

907642

Sigma-Aldrich

Cuprous thiocyanate

greener alternative

35 mg/mL in diethyl sulfide

Synonym(s):

Copper thiocyanate perovskite hole transport material, Copper(I) thiocyanate perovskite hole transport material, Thiocyanic acid in diethyl sulfide

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$374.00

$374.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$374.00

About This Item

Linear Formula:
CuSCN
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

$374.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

[Cu]SC#N

InChI

1S/CHNS.Cu/c2-1-3;/h3H;/q;+1/p-1

InChI key

PDZKZMQQDCHTNF-UHFFFAOYSA-M

General description

We are committed to bringing you Greener Alternative Products,which adhere to one or more of The 12 Principles of Greener Chemistry. This product is an enabling product used as a Hole Transport Material for high-performance solar cells and thus has been enhanced for energy efficiency. Click here for more information.

Application

Layers of CuSCN exhibit high hole mobility, high thermal stability, and exceptional optical transparency across the UV-Vis/NIR spectrum. CuSCN finds application as an alternative to organic hole transport materials such as PEDOT:PSS and Spiro-MeOTAD in perovskite-based solar cells (PSCs).[1][2][3][4] Photovoltaic conversion efficiencies of >20% have been reported in PSCs with CuSCN [5]. In addition, compared to Spiro-OMeTAD the CuSCN-based PSCs have been shown to exhibit better thermal stability.[5]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

25.7 °F - closed cup

Flash Point(C)

-3.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Planar perovskite solar cells employing copper(I) thiocyanate/N,N′-di(1-naphthyl)-N,N′-diphenyl-(1,1′-biphenyl)-4,4′-diamine bilayer structure as hole transport layers
Tseng Z L ,et al.
Japanese Journal of Applied Physics, 57, 02CE07-02CE07 (2018)
High-Efficiency Organic Photovoltaic Cells Based on the Solution-Processable Hole Transporting Interlayer Copper Thiocyanate (CuSCN) as a Replacement for PEDOT:PSS
Gross N Y, et al.
Advanced Energy Materials, 5, 1401529-1401529 (2015)
Perovskite solar cells with CuSCN hole extraction layers yield stabilized efficiencies greater than 20%
N Arora, et al.
Science, 358(6364), 768-771 (2017)
Pichaya Pattanasattayavong et al.
Chemical communications (Cambridge, England), 49(39), 4154-4156 (2012-12-12)
The optical, structural and charge transport properties of solution-processed films of copper(I) thiocyanate (CuSCN) are investigated in this work. As-processed CuSCN films of ~20 nm in thickness are found to be nano-crystalline, highly transparent and exhibit intrinsic hole transporting characteristics
Pichaya Pattanasattayavong et al.
Advanced materials (Deerfield Beach, Fla.), 25(10), 1504-1509 (2013-01-03)
The wide bandgap and highly transparent inorganic compound copper(I) thiocyanate (CuSCN) is used for the first time to fabricate p-type thin-film transistors processed from solution at room temperature. By combining CuSCN with the high-k relaxor ferroelectric polymeric dielectric P(VDF-TrFE-CFE), we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service