Merck
  • Electric field-induced hole transport in copper(I) thiocyanate (CuSCN) thin-films processed from solution at room temperature.
Electric field-induced hole transport in copper(I) thiocyanate (CuSCN) thin-films processed from solution at room temperature.
Chemical communications (Cambridge, England) (2012-12-12)
Pichaya Pattanasattayavong, Guy Olivier Ngongang Ndjawa, Kui Zhao, Kang Wei Chou, Nir Yaacobi-Gross, Brian C O'Regan, Aram Amassian, Thomas D Anthopoulos
ABSTRACT
The optical, structural and charge transport properties of solution-processed films of copper(I) thiocyanate (CuSCN) are investigated in this work. As-processed CuSCN films of ~20 nm in thickness are found to be nano-crystalline, highly transparent and exhibit intrinsic hole transporting characteristics with a maximum field-effect mobility in the range of 0.01-0.1 cm(2) V(-1) s(-1).
Sigma-Aldrich
Cuprous thiocyanate, 35 mg/mL in diethyl sulfide