Skip to Content
Merck
All Photos(1)

Key Documents

901974

Sigma-Aldrich

SunTronic® UV curing jettable insulator for low-K dielectric applications

greener alternative

Synonym(s):

Inkjet UV insulator, SunTronic® U6415 JET9525 clear dielectric, UV curing insulator coating, dielectric ink

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$309.00

$309.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$309.00

About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

$309.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

Cure dose required (un-doped mercury): 100-300 mJ/cm2
Jetting temperature: 50°C

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface tension

32-37 mN/m

viscosity

11-13 cP (at jetting temperature)

greener alternative category

storage temp.

2-8°C

General description

SunTronic jettable insulator is a UV-curing, inkjet printable insulator coating,suitable for low-K dielectric applications, printed electronics and related applications.
The insulator can be jetted through various piezo DOD inkjet printheads, although full printhead compatibility will need to be verified in each case. The product can be cured using an un-doped medium pressure mercury source. If over printing the dielectric with a SunTronic jettable silver ink then a cure dose of 300 mJ/cm2 is recommended.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Click here for more information.

Legal Information

SunTronic is a registered trademark of Sun Chemical, Inc.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 - STOT SE 3

Target Organs

Liver,Upper respiratory tract, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

204.8 °F

Flash Point(C)

96 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Questions

  1. What chemicals (besides acetone, isopropyl) are necessary for cleaning this ink from the nozzles and pipes of the inkjet system?

    1 answer
    1. For a larger printer, an ester solvent, such as Carbitol Acetate, is recommended. This is primarily due to the compatibility with most industrial print-heads. However, this assumes that the ink has not be cured. If the ink is cured, ultrasonic cleaning and back flushing may remove the cured ink. Any light source is to be avoided at all times to prevent curing of the ink in and around the print-head.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service