Skip to Content
Merck
All Photos(2)

Key Documents

736465

Sigma-Aldrich

Silver, dispersion

nanoparticle, 30-35 wt. % in triethylene glycol monoethyl ether, spec. resistivity 11 μΩ-cm, for printing on plastic films

Synonym(s):

Silverjet DGP-40LT-15C, silver ink

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$1,480.00
100 G
$4,580.00

$1,480.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$1,480.00
100 G
$4,580.00

About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$1,480.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

description

surface tension 35 - 40 dyn/cm

form

dispersion
nanoparticle

concentration

30-35 wt. % in triethylene glycol monoethyl ether

refractive index

n20/D 1.333

particle size

≤50 nm

viscosity

10-18 cP

density

1.45 g/mL±0.05 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver is a good conductor and is resistant to oxidation. It is used in printed conductors and printable inks (based on silver nanoparticles).Other large scale applications of nanosilver include metallization of solar cells, thin film electronics, sensors, and catalysts.

Curing Temperature: 100-150 °C

Recommended substrates: Plastic films

Recommended washing solvent: Polar Solvents: Ethyl Alcohol, IPA etc.

Application

  • Enhanced Adsorption of Trace Ethylene on Ag/NZ5 Modified with Ammonia: Hierarchical Structure and Metal Dispersion Effects.: This study examines the effects of hierarchical structure and metal dispersion in enhancing the adsorption of trace ethylene using silver/nanozeolite 5 modified with ammonia, exploring potential industrial applications for improved air quality and storage conditions (Qi et al., 2024).
  • Structure-Activity Relationships between the State of Silver on Different Supports and Their I(2) and CH(3)I Adsorption Properties.: The paper details the correlation between the physical state of silver dispersed on various supports and its properties in adsorbing iodine and methyl iodide, which could be crucial for developing better filtration and purification systems (Azambre et al., 2021).
  • New Relevant Descriptor of Linear QNAR Models for Toxicity Assessment of Silver Nanoparticles.: This publication introduces a new quantitative nanostructure-activity relationship (QNAR) descriptor that enhances the toxicity assessment of silver nanoparticles, significant for pharmaceutical applications where safety is paramount (Kudrinskiy et al., 2020).
  • Resolving the puzzle of single-atom silver dispersion on nanosized γ-Al(2)O(3) surface for high catalytic performance.: Investigates the single-atom dispersion of silver on nano-sized γ-alumina surfaces, providing insights into optimizing the catalytic efficiency and potential applications in various chemical industries (Wang et al., 2020).

Analysis Note

Our Silver dispersion contains 30-35 wt% of silver nanoparticles (measured after dry oven baking @250 Deg C for 3 hours), providing a spec. resistivity of 11 μΩ-cm.

Legal Information

Product of ANP, Inc.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Inkjet printing is one of the key enabling technologies of printed electronics. Inkjet printing technology classification, aspects of materials (inks, substrates) and respective pre-and post-processing steps are discussed.

The ability to pattern conductive electrodes is technologically relevant for several applications, including photovolatics, displays, sensors, and biomedical devices.

Functional materials for printed electronics applications enable flexible displays, RFID tags, and biomedical sensors.

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service