Skip to Content
Merck
All Photos(1)

Key Documents

901267

Sigma-Aldrich

Hexamethylborazine

≥95%

Synonym(s):

1,2,3,4,5,6-Hexamethyl-1,3,5,2,4,6-triazatriborinane

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$210.00
1 G
$506.00

$210.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$210.00
1 G
$506.00

About This Item

Empirical Formula (Hill Notation):
C6H18B3N3
CAS Number:
Molecular Weight:
164.66
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

$210.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder or crystals

reaction suitability

core: boron
reagent type: catalyst

mp

99 °C

storage temp.

−20°C

SMILES string

B1(N(B(N(B(N1C)C)C)C)C)C

InChI

1S/C6H18B3N3/c1-7-10(4)8(2)12(6)9(3)11(7)5/h1-6H3

InChI key

LCHQMXUQYONIOI-UHFFFAOYSA-N

Application

Hexamethylborazine has been developed by the Szymczak lab[1] for the delivery of alkyl and hydride nucleophiles[2] in addition to being a key component in the synthesis of recyclable trifluoromethylation reagents.[3][4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manganese-mediated hydride delivery to a borazine by stepwise reduction and protonation.
Carter TJ, et al.
Organometallics, 33 (7), 1540-1543 (2014)
Anna S Lisovenko et al.
Inorganic chemistry, 49(22), 10357-10369 (2010-10-23)
Donor-acceptor complexes of borazine (BZ) and its substituted derivatives with Lewis acids (A = MCl(3), MBr(3); M = B, Al, Ga) and Lewis bases (D = NH(3), Py) have been theoretically studied at the B3LYP/TZVP level of theory. The calculations
Recyclable Trifluoromethylation Reagents from Fluoroform.
Geri J B and Szymczak N K
Journal of the American Chemical Society, 139(29), 9811-9814 (2017)
Borazine?CF3? Adducts for Rapid, Room Temperature, and Broad Scope Trifluoromethylation.
Geri J B, et al.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 130(5), 1395-1399 (2018)
Tyler J Carter et al.
Angewandte Chemie (International ed. in English), 51(52), 13168-13172 (2012-11-20)
Recharging spent BN fuel: {Cr(CO)(3)} mediates the reduction of borazines by hydride and methyl nucleophiles to generate anionic complexes of dearomatized hexamethylborazine. Subsequent quenching leads to the release of a substituted cyclotriborazane, successfully demonstrating the stepwise reduction of a B=N

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service