Skip to Content
Merck
All Photos(1)

Key Documents

184225

Sigma-Aldrich

Hydrogen fluoride pyridine

pyridine ~30 %, hydrogen fluoride ~70 %

Synonym(s):

HF-Pyridine, Olah′s reagent, PPHF, Poly(pyridine fluoride), Pyridine hydrofluoride, Pyridinium poly(hydrogen fluoride), Pyridinium polybifluoride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$104.00
100 G
$186.00

$104.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$104.00
100 G
$186.00

About This Item

CAS Number:
Beilstein:
4750151
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22

$104.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

composition

hydrogen fluoride, ~70%
pyridine, ~30%

density

1.1 g/mL at 20 °C (lit.)

storage temp.

−20°C

SMILES string

F[H].c1ccncc1

InChI

1S/C5H5N.FH/c1-2-4-6-5-3-1;/h1-5H;1H

InChI key

GRJJQCWNZGRKAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

May contain or form low levels of calcium fluoride during storage. Presence of this does not impact the specification values.

Application

Convenient form of anhydrous HF, stable up to 50°C. Has been used for the preparation of β-fluoroamines from amino alcohols and for the fluorination of acetylenes.[1][2]
Used together with hypervalent iodine(III) reagents for ipso-fluorination of para-substituted phenols providing cyclohexadienones.[3] Employed with Selectfluor® (Catalog No. 439479) for geminal fluorination of 2,2-diaryl-1,3-dithiolanes.[4]
Reactant for preparation of:
  • Epimers of shikimic acid with the features of fucosylated glycans via zinc-mediated reductive ring opening followed by a Barbier reaction
  • Vaccinia H1-related (VHR) phosphatase inhibitor with a nonacidic phosphate-mimicking core structure
  • Candidates for nucleic acid drugs
  • ω-substituted gem-difluoroalkanes by oxidative desulfurization-difluorination

Reagent for:
  • Halofluorination reactions

Legal Information

Selectfluor is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Journal of Fluorine Chemistry, 37, 343-343 (1987)
Tetrahedron, 60, 6629-6629 (2004)
Tetrahedron Letters, 32, 69-69 (1991)
V Prakash Reddy et al.
Chemical communications (Cambridge, England), (5), 654-656 (2005-01-27)
2,2-diaryl-1,3-dithiolanes, readily obtainable from diaryl ketones, were transformed into the corresponding gem-difluoro compounds using a novel reagent combination involving Selectfluor and pyridinium polyhydrogen fluoride (PPHF) under mild conditions in moderate to good yields.
Hui Shen et al.
Infection and immunity, 83(7), 2694-2704 (2015-04-22)
Fungi can shield surface pathogen-associated molecular patterns (PAMPs) for evading host immune attack. The most common and opportunistic human pathogen, Candida albicans, can shield β-(1 3)-glucan on the cell wall, one of the major PAMPs, to avoid host phagocyte Dectin-1

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service