Skip to Content
Merck
All Photos(1)

Key Documents

901158

Sigma-Aldrich

2-(2-(2-Methoxyethoxy)ethoxy) ethanamine

≥95%

Synonym(s):

Tri(ethylene glycol) methyl ether amine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$429.00

$429.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$429.00

About This Item

Empirical Formula (Hill Notation):
C7H17NO3
CAS Number:
Molecular Weight:
163.21
EC Number:
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

$429.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

liquid

refractive index

n/D 1.441

density

0.987

storage temp.

2-8°C

SMILES string

NCCOCCOCCOC

InChI

1S/C7H17NO3/c1-9-4-5-11-7-6-10-3-2-8/h2-8H2,1H3

InChI key

OKUWOEKJQRUMBW-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Choosing the right crosslinker for your biochemical or bioconjugation research can be challenging. Our selection guide can help you find the perfect match for your applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service