Skip to Content
Merck
All Photos(1)

Key Documents

901154

Sigma-Aldrich

2-(2-(2-Ethoxyethoxy)ethoxy)ethanamine

Synonym(s):

3,6,9-Trioxaundecylamine, N-[2-(2-(2-Ethoxyethoxy)ethoxy)ethyl]amine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$375.00

$375.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$375.00

About This Item

Empirical Formula (Hill Notation):
C8H19NO3
CAS Number:
Molecular Weight:
177.24
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

$375.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

density

0.974

SMILES string

NCCOCCOCCOCC

InChI

1S/C8H19NO3/c1-2-10-5-6-12-8-7-11-4-3-9/h2-9H2,1H3

InChI key

WWJVRDMJNJTOBL-UHFFFAOYSA-N

Application

2-(2-(2-Ethoxyethoxy)ethoxy)ethanamine can be used as a reactant to synthesize:
  • 2-(2-(2-(2-Ethoxyethoxy)ethoxy)ethyl)-4-nitroisoindoline-1,3-dione, a key intermediate used to prepare excited-state intramolecular proton transfer (ESIPT)-based fluorescent probe for the detection of palladium.[1]
  • Acetamidine derivatives by condensation reaction with N,N-dimethylacetamide dimethyl acetal.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A water soluble ESIPT-based fluorescent chemodosimeter for the ratiometric detection of palladium ions in aqueous solution and its application in live-cell imaging
Huang J, et al.
New. J. Chem., 42(19), 15587-15592 (2018)
Jitendra R Harjani et al.
The Journal of organic chemistry, 76(6), 1683-1691 (2011-02-15)
The condensation of primary amine with N,N-dimethylacetamide dimethyl acetal yields a mixture of acetamidine and imidate ester. The product distribution in this reaction depends on the temperature, solvent, and structure of the primary amine. It is possible to suppress the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service