Skip to Content
Merck
All Photos(1)

Documents

QBD10175

Sigma-Aldrich

m-dPEG®4-amine

Synonym(s):

2-[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]ethylamine, 3,6,9,12-Tetraoxatridecylamine, Polyethylene glycol, Triethylene glycol 2-aminoethyl methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H21NO4
CAS Number:
Molecular Weight:
207.27
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: chemical modification reagent
reagent type: cross-linking reagent
reactivity: carboxyl reactive

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

NCCOCCOCCOCCOC

InChI

1S/C9H21NO4/c1-11-4-5-13-8-9-14-7-6-12-3-2-10/h2-10H2,1H3

InChI key

DQTQYVYXIOQYGN-UHFFFAOYSA-N

Features and Benefits

m-dPEG®4-amine provides a variety of means to modify small molecules, peptides, proteins, and functionalized surfaces (metals, glass, and others). The single molecular weight methoxy-terminated dPEG® molecules are hydrophilic and non-immunogenic of exact length. There are four repeat units in this linear dPEG® chain. Modification with m-dPEG® reagent can improve water solubility, increase hydrodynamic volume, eliminate or reduce non-specific binding, protect peptides and proteins from proteolysis, reduce or eliminate immunogenicity, and increase serum half-life in vivo. The m-dPEG®4-amine reacts with carboxylic acids and active esters.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service