Skip to Content
Merck
All Photos(3)

Key Documents

900713

Sigma-Aldrich

Graphene quantum dots

aqua green luminescent, powder

Synonym(s):

GQDs

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$177.00

$177.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$177.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
11101522
NACRES:
NA.23

$177.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

description

topographic height: 1–2.0 nm

form

powder

diameter

<5 nm

fluorescence

λex 485 nm; λem 525 nm FWHM 70 nm, quantum yield >17%

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Graphene quantum dots (GQDs), sheets of few-layered graphene and lateral dimensions smaller than 100 nm, possess strong quantum confinement and edge effects. Thus, they possess unique physical properties such as strong photoluminescence, which can be tailored for specific applications by controlling their size, shape, defects, and functionality.
In contrast to classic QDs, such as metal or silicon quantum dots, GQDs are biocompatible, photo-stable, and inherit superior thermal, electrical, and mechanical properties from the graphene. These features can greatly contribute to various state-of-the-art applications:

  • Optical brighteners.
  • Taggants for security application.
  • Bioimaging markers.
  • Fluorescent polymers.
  • Antibacterial, antibiofouling, disinfection systems.
  • Heavy metals, humidity and pressure sensors.
  • Batteries.
  • Flash memory devices.
  • Photovoltaic devices.
  • Light-emitting diodes.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianhua Shen et al.
Chemical communications (Cambridge, England), 48(31), 3686-3699 (2012-03-14)
Similar to the popular older cousins, luminescent carbon dots (C-dots), graphene quantum dots or graphene quantum discs (GQDs) have generated enormous excitement because of their superiority in chemical inertness, biocompatibility and low toxicity. Besides, GQDs, consisting of a single atomic
An electrochemical avenue to green-luminescent graphene quantum dots as potential electron-acceptors for photovoltaics.
Yan Li et al.
Advanced materials (Deerfield Beach, Fla.), 23(6), 776-780 (2011-02-03)

Articles

Properties, synthesis & applications of Graphene Quantum Dots explained. GQDs have advantages of nontoxicity, good solubility, stable photoluminescence & better surface grafting.

Graphene nanoribbons (GNRs) are quasi-one-dimensional narrow strips of graphene comprised of sp2-hybridized carbon atoms arranged into hexagonal honeycomb lattice configurations.

Perovskite quantum dots research progresses overcoming challenges, enabling rapid development of light-emitting devices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service