Skip to Content
Merck
All Photos(1)

Key Documents

777935

Sigma-Aldrich

CdTe core-type quantum dots

COOH functionalized, fluorescence λem 520 nm, powder

Synonym(s):

Fluorescent nanocrystals, QDs, artificial atoms

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$581.00
25 MG
$1,270.00

$581.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$581.00
25 MG
$1,270.00

About This Item

Linear Formula:
CdTe
EC Number:
UNSPSC Code:
26111700
NACRES:
NA.23

$581.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

fluorescence

λem 520 nm
, quantum yield 36%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Cadmium Telluride Quantum Dots (CdTe QDs) are available in solid form. They are coated with carboxyl groups which makes them water soluble to make aqueous solutions. These carboxyl functionalized hydrophilic quantum dots can be further conjugated for use as labels. CdTe QDs can be used for optical and display applications; solar cells and biomedical applications.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julie Hottechamps et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 21(9), 853-862 (2020-02-23)
As luminescent quantum dots (QDs) are known to aggregate themselves through their chemical activation by carbodiimide chemistry and their functionalization with biotin molecules, we investigate both effects on the fluorescence properties of CdTe QDs and their impact on Förster Resonant
P Haro-González et al.
Small (Weinheim an der Bergstrasse, Germany), 8(17), 2652-2658 (2012-06-16)
The potential use of CdTe quantum dots as luminescence nano-probes for lifetime fluorescence nano-thermometry is demonstrated. The maximum thermal sensitivity achievable is strongly dependent on the quantum dot size. For the smallest sizes (close to 1 nm) the lifetime thermal
R S Aga et al.
Nanotechnology, 20(46), 465204-465204 (2009-10-23)
A photosensitized high-surface area transparent electrode has been employed to increase the short circuit current of a photovoltaic device with a blend of poly(3-hexylthiophene) (P3HT) and (6,6)-phenyl C61 butyric acid methyl ester (PCBM) as the active layer. This is achieved
A Rameshkumar et al.
Colloids and surfaces. B, Biointerfaces, 101, 74-82 (2012-07-17)
This study was undertaken to evaluate the antioxidant potential of an aqueous extract from Merremia emarginata leaves because this plant has a very high flavonoid and phenol content. The in vitro antioxidant activity was measured by diphenyl-1-picrylhydrazyl (DPPH), 2,2'-azino-bis (3-ethylbenzthiazoline-6-sulphonic

Articles

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

Professor Sharma and colleagues review the synthesis and applications of this novel material. This includes a discussion of the unique properties of quantum dots and their suitability for solar cell applications, along with common synthesis techniques used to develop these materials.

Professor Xiaohu Gao (University of Washington, USA) provides a overview of recent quantum dot (QD) advancements and their potential for advancing bioassay and bioimaging technologies.

Perovskite quantum dots research progresses overcoming challenges, enabling rapid development of light-emitting devices.

See All

Protocols

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Questions

1–3 of 3 Questions  
  1. I would like to know the TEM size of CdTe core-type quantum dots COOH functionalized (777935), fluorescence λem 520 nm, powder for calculation purpose

    1 answer
    1. The particle size of this product is approximately 2.0 nm. Unfortunately, there is no information on how the particle size is measured.

      Helpful?

  2. What is the molecular weight of 777935 - CdTe core-type quantum dots COOH functionalized, fluorescence λem 520 nm, powder

    1 answer
    1. The molecular weight of this product is approximately 16000 Da.

      Helpful?

  3. CdTe core-type quantum dots (COOH functionalized, fluorescence λem 520 nm, powder) Hi,I would like to ask about the concentration used of the Q.D and the amino functionalized DNA for EDC conjugation. EDC concentration,Q.D. concentration, DNA conc.

    1 answer
    1. This product is not tested for suitability in any specific application. While these Quantum Dots are hydrophilic, the researcher would have to determine suitability. The links below may offer some helpful information:
      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC5071820/
      https://pubs.acs.org/doi/10.1021/nn1021346

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service