Skip to Content
Merck
All Photos(1)

Key Documents

900214

Sigma-Aldrich

CdSe/ZnS core-shell type quantum dots

stabilized with octadecylamine ligands, fluorescence λem 520 nm, 5 mg/mL in toluene

Synonym(s):

Fluorescent nanocrystals, QDs

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$117.00

$117.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$117.00

About This Item

Linear Formula:
CdSe/ZnS
UNSPSC Code:
26111700
NACRES:
NA.23

$117.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

dispersion

concentration

5 mg/mL in toluene

λmax

505 nm

fluorescence

λem 520 nm
λem 520 nm±10 nm, quantum yield >50%

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Reactive group: Octadecylamine.

Application

Our highly purified CdSe/ZnS quantum dots with an octadecylamine surface contain extremely low free organic ligands and impurities and are well suited for display, solid state lighting and solar cell applications. For example, they can be used as electroluminescence emitter in quantum dots based LED′s or can be embedded in polymer matrix as the backlight of LCD display. CdSe/ZnS quantum dots in solid form can be dispersed in most non-polar organic solvents such as toluene, chloroform, hexane etc.[1][2][3]

Legal Information

Product of Ocean Nanotech, LLC.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3

Flash Point(F)

39.2 °F

Flash Point(C)

4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingjiang Sun et al.
ACS nano, 3(3), 737-743 (2009-03-25)
Owing to their narrow bright emission band, broad size-tunable emission wavelength, superior photostability, and excellent flexible-substrate compatibility, light-emitting diodes based on quantum dots (QD-LEDs) are currently under intensive research and development for multiple consumer applications including flat-panel displays and flat
Bright, multicoloured light-emitting diodes based on quantum dots.
Sun Q, et al.
Nature Photonics, 1, 717-722 (2007)
Zhanao Tan et al.
Nano letters, 7(12), 3803-3807 (2007-11-03)
We report a multilayer solution-processed blue light-emitting diode based on colloidal core/shell CdS/ZnS nanocrystal quantum dots (QDs). At a low-operating voltage of 5.5 V, the device emits spectrally pure blue radiation at 460 nm with a narrow full-width-at-half-maximum bandwidth of

Articles

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Questions

  1. What are the dimensions of the CdSe particle inside the ZnS shell for product 900214?

    1 answer
    1. The CdSe core size of the this product (900214) is about 2.8 nm.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service