Skip to Content
Merck
All Photos(2)

Documents

900708

Sigma-Aldrich

Graphene quantum dots

blue luminescent, 1 mg/mL in H2O

Synonym(s):

GQD

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200

description

topographic height: 1-2.0 nm

form

suspension

concentration

1 mg/mL in H2O

color

colorless

diameter

<5 nm

fluorescence

λex 350 nm; λem 445 nm FWHM 65 nm, quantum yield >65%

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Graphene quantum dots (GQDs), sheets of few-layered graphene and lateral dimensions smaller than 100 nm, possess strong quantum confinement and edge effects. Thus, they possess unique physical properties such as strong photoluminescence, which can be tailored for specific applications by controlling their size, shape, defects, and functionality. In contrast to classic QDs, such as metal or silicon quantum dots, GQDs are biocompatible, photo-stable, and inherit superior thermal, electrical, and mechanical properties from the graphene. These features can greatly contribute to various state-of-the-art applications:
  • Optical brighteners.
  • Taggants for security application.
  • Bio-imaging markers.
  • Fluorescent polymers.
  • Antibacterial, antibiofouling, disinfection systems.
  • Heavy metals, humidity and pressure sensors.
  • Batteries.
  • Flash memory devices.
  • Photovoltaic devices.
  • Light-emitting diodes.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Cristina Stefanov et al.
Mikrochimica acta, 187(11), 611-611 (2020-10-17)
A gold nanoparticle/graphene quantum dots (AuNP/GQD) nanozyme-modified screen-printed carbon electrode (SPCE) was developed for fast and ultrasensitive determination of quercetin by square-wave voltammetry. The nanosensing device was inserted in miniaturized equipment. Under the optimal experimental conditions, a good linear relationship between
Jianhua Shen et al.
Chemical communications (Cambridge, England), 48(31), 3686-3699 (2012-03-14)
Similar to the popular older cousins, luminescent carbon dots (C-dots), graphene quantum dots or graphene quantum discs (GQDs) have generated enormous excitement because of their superiority in chemical inertness, biocompatibility and low toxicity. Besides, GQDs, consisting of a single atomic
An electrochemical avenue to green-luminescent graphene quantum dots as potential electron-acceptors for photovoltaics.
Yan Li et al.
Advanced materials (Deerfield Beach, Fla.), 23(6), 776-780 (2011-02-03)

Articles

Properties, synthesis & applications of Graphene Quantum Dots explained. GQDs have advantages of nontoxicity, good solubility, stable photoluminescence & better surface grafting.

Graphene nanoribbons (GNRs) are quasi-one-dimensional narrow strips of graphene comprised of sp2-hybridized carbon atoms arranged into hexagonal honeycomb lattice configurations.

Perovskite quantum dots research progresses overcoming challenges, enabling rapid development of light-emitting devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service