Skip to Content
Merck
All Photos(1)

Key Documents

900313

Sigma-Aldrich

Poly(caprolactone-co-glycolide)

caprolactone:glycolide 45:55, viscosity 1.5 dL/g 

Synonym(s):

PCL-PGA

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$449.00

$449.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$449.00

About This Item

Linear Formula:
CH3(C6H10O2)m(C4H4O4)nCH3
UNSPSC Code:
12162002
NACRES:
NA.23

$449.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

chunks

Quality Level

feed ratio

caprolactone:glycolide 45:55

viscosity

1.5 dL/g

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service