Skip to Content
Merck
All Photos(1)

Key Documents

747009

Sigma-Aldrich

Poly(styrene)-block-poly(acrylic acid)

PS:PAA 30,000:2,000, PDI ≤1.1

Synonym(s):

PS-PAA, PS-PAA amphiphilic diblock copolymer

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$598.00

$598.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$598.00

About This Item

Linear Formula:
C4H6N(C8H8)x(C3H4O2)yH
UNSPSC Code:
12352100
NACRES:
NA.23

$598.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

form

powder

mol wt

Mn 1,000-2,000 (poly(acrylic acid))
Mn 27,000-31,000 (polystyrene)
Mn 28,000-33,000 (total)

PDI

≤1.1

Application

Polystyrene-block-polyacrylic acid is a diblock copolymer used for making polymeric micelles/ vesicles (polymersomes) and other encapsulation applications[1], [2]. This PS-block-PAA copolymer is 10 wt. % PAA; and should form micelles in water in the 100~200 nm range. The polystyrene degree of polymerization (DP) is 275 and the polyacrylic acid DP is 30.

Legal Information

Sold for research purposes only and subject to field restriction: see www.sigmaaldrich.com/raftlicense. Patents: 7,714,075; 7,250,479; 7,666,962; 6,642,318; 6,747,111; W2010/8356.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaoutar Moumile et al.
Journal of clinical microbiology, 42(2), 923-924 (2004-02-10)
We describe a case of primary purulent culture-negative pericarditis caused by Neisseria meningitidis serogroup C occurring in an 8-month-old previously healthy boy, which was detected in pericardial fluid by broad-spectrum PCR amplification.
Dennis E Discher et al.
Science (New York, N.Y.), 297(5583), 967-973 (2002-08-10)
Vesicles are microscopic sacs that enclose a volume with a molecularly thin membrane. The membranes are generally self-directed assemblies of amphiphilic molecules with a dual hydrophilic-hydrophobic character. Biological amphiphiles form vesicles central to cell function and are principally lipids of

Articles

Reversible addition–fragmentation chain transfer (RAFT) polymerization is rapidly moving to the forefront in construction of drug and gene delivery vehicles.

The development of drugs that target specific locations within the human body remains one of the greatest challenges in biomedicine today.

Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service