Skip to Content
Merck
All Photos(1)

Key Documents

901011

Sigma-Aldrich

Poly(caprolactone-co-glycolide)

40:60, viscosity 1.6 dL/g 

Synonym(s):

PGA-PCL, PGCL

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$449.00

$449.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$449.00

About This Item

Linear Formula:
(C4H4O4)m(C6H10O2)n
UNSPSC Code:
12162002
NACRES:
NA.23

$449.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

chips

feed ratio

glycolide:caprolactone 60:40±5

color

light yellow to tan

viscosity

1.2-2.0 dL/g
1.6 dL/g

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Poly(caprolactone-co-glycolide) (PGCL) is a biodegradable and biocompatible copolymer synthesized from the ring opening polymerization of glycolide and ε-caprolactone. Due to the ester linkages found in the polymer backbone, these copolymers are able to degrade via hydrolysis. Depending on the ratio of monomers used and polymer block structure, different forms of PGCL can be obtained. These differing forms yield materials with different degrees of crystallinity, thermal transitions, solubility, and rates of degradation.

Application

Poly(caprolactone-co-glycolide) (PGCL) is a biodegradable and biocompatible copolymer for use in biomedical research devices, such as implants, sutures, prosthetic devices, scaffolds for tissue engineering applications, micro and nanoparticles. PGA-PCL is also utilized in drug delivery research, such as in the synthesis and application of polymer-drug conjugates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Period of biodegradation? Glass transition temperature?

    1 answer
    1. The period of biodegradation has not been determined. The glass transition temperature (Tg) is approximately 2.86°C and the melting temperature (Tm) is approximately 160°C.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service