Skip to Content
Merck
All Photos(1)

Key Documents

900075

Sigma-Aldrich

Vitamin K2 (MK-7)-4′,5,6,7,8,8′-13C6

≥99 atom % 13C, ≥95% (CP)

Synonym(s):

Menaquinone-7-4′,5,6,7,8,8′-13C6

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$7,880.00

$7,880.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 MG
$7,880.00

About This Item

Empirical Formula (Hill Notation):
13C6C40H64O2
Molecular Weight:
654.96
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

$7,880.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

≥99 atom % 13C

Quality Level

Assay

≥95% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

color

yellow

mass shift

M+6

storage temp.

−20°C

SMILES string

O=C1C(C)=C(C/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CCC=C(C)C)C([13C]2=[13C][13C]=[13C][13C]=[13C]21)=O

InChI

1S/C46H60O2/c1-34(2)18-12-19-35(3)20-13-21-36(4)22-14-23-37(5)24-15-25-38(6)26-16-27-39(7)28-17-29-40(8)32-33-42-41(9)45(47)43-30-10-11-31-44(43)46(42)48/h18,20,22,24,26,28,32H,12-17,19,21,23,25,27,29,33H2,1-9H3/b35-20+,36-22+,37-24+,38-26+,39-28+,40-32+/i10+1,11+1,30+1,31+1,43+1,44+1

InChI key

QNFXTFFPNJWCQL-MUMAHXDWSA-N

Related Categories

General description

Vitamin K2, also known as menaquinone, is a polyisoprenoid-substituted napthoquinone. Menaquinone-7 (MK-7) is a subtype of vitamin K2. It is found in animals, soy protein, fermented products and is primarily produced by bacteria.[1][2]
Vitamin K2 (MK-7)-4′,5,6,7,8,8′-13C6 is a vitamin K2 isotope wherein C-4′, C-5, C-6, C-7, C-8, and C-8′ carbons are replaced by 13C6.

Application

Vitamin K2 (MK-7)-4′,5,6,7,8,8′-13C6 can be used as a stable isotope internal standard for data interpretation in specific biological samples.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitamin K
Textbook of Veterinary Physiological Chemistry (2014)
Production of menaquinone (vitamin K2)-7 by Bacillus subtilis
Sato T, et al.
Journal of Bioscience and Bioengineering, 91(1), 16-20 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service