Skip to Content
Merck
All Photos(1)

Key Documents

1381119

USP

Menaquinone-7

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Vitamin K2-(MK-7), (all-E)–2-(3,7,11,15,19,23,27-Heptamethyl-2,6,10,14,18,22,26-octacosaheptaenyl)-3-methyl-1,4-naphthalenedione

Sign Into View Organizational & Contract Pricing

Select a Size

3 X 50 MG
$557.00

$557.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
3 X 50 MG
$557.00

About This Item

Empirical Formula (Hill Notation):
C46H64O2
CAS Number:
Molecular Weight:
649.00
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$557.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

menaquinone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

O=C1c2c(cccc2)C(=O)C(=C1C\C=C(\CC\C=C(\CC\C=C(\CC\C=C(\CC\C=C(\CC\C=C(\CCC=C(C)C)/C)/C)/C)/C)/C)/C)C

InChI

1S/C46H64O2/c1-34(2)18-12-19-35(3)20-13-21-36(4)22-14-23-37(5)24-15-25-38(6)26-16-27-39(7)28-17-29-40(8)32-33-42-41(9)45(47)43-30-10-11-31-44(43)46(42)48/h10-11,18,20,22,24,26,28,30-32H,12-17,19,21,23,25,27,29,33H2,1-9H3/b35-20+,36-22+,37-24+,38-26+,39-28+,40-32+

InChI key

RAKQPZMEYJZGPI-LJWNYQGCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.

Application

This USP reference standard is intended for use in specified quality tests and assays.
Also used to prepare standard solutions as described in the following United States Pharmacopeia (USP) monographs:
  • Menaquinone-7
  • Bacillus subtilis subsp. subtilis Menaquinone-7 Extract
  • Menaquinone-7 Tablets
  • Menaquinone-7 Capsules

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Disclaimer

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bacillus subtilis subsp. subtilis Menaquinone-7 Extract
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(3), 5152-5152 (2015)
Menaquinone-7
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(4), 5146-5146 (2020)
Menaquinone-7 Tablets
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(2), 5151-5151 (2015)
Menaquinone-7 Capsules
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(2), 5147-5147 (2015)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service