Skip to Content
Merck
All Photos(1)

Key Documents

809888

Sigma-Aldrich

Vitamin K1-4a,5,6,7,8,8a-13C6

≥99 atom % 13C, ≥98% (CP)

Synonym(s):

Phylloquinone-13C6

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$3,390.00

$3,390.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request more information

Select a Size

Change View
1 MG
$3,390.00

About This Item

Empirical Formula (Hill Notation):
13C6C25H46O2
Molecular Weight:
456.65
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

$3,390.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request more information

isotopic purity

≥99 atom % 13C

Quality Level

Assay

≥98% (CP)

form

liquid

technique(s)

mass spectrometry (MS): suitable

color

yellow to orange

mp

-20 °C

mass shift

M+6

storage temp.

−20°C

SMILES string

CC(C)CCCC(C)CCCC(C)CCC/C(C)=C/CC1=C(C)C([13C]2=[13CH][13CH]=[13CH][13CH]=[13C]2C1=O)=O

InChI

1S/C31H46O2/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-17-25(5)20-21-27-26(6)30(32)28-18-7-8-19-29(28)31(27)33/h7-8,18-20,22-24H,9-17,21H2,1-6H3/b25-20+/i7+1,8+1,18+1,19+1,28+1,29+1

InChI key

MBWXNTAXLNYFJB-HDMSKXMFSA-N

Related Categories

General description

Vitamin K1, also known as phylloquinone, is a fat-soluble vitamin found in algae, cyanobacteria, and plants. It is involved in the synthesis of some important proteins having applications in blood clotting and bone metabolism.[1][2]
Vitamin K1-4a,5,6,7,8,8a-13C6 is an isotope of vitamin K1 wherein C-4a, C-5, C-6, C-7, C-8, C-8a carbons are replaced by 13C6 carbon.

Application

Vitamin K1-4a,5,6,7,8,8a-13C6 can be used as a stable isotope internal standard for accurate data interpretation in specific biological samples.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitamin K1 supplementation to improve the stability of anticoagulation therapy with vitamin K antagonists: a dose-finding study
Gebuis EPA, et al.
Haematologica, 96(4), 583-589 (2011)
Phylloquinone (vitamin K1) content of vegetables
Damon M, et al.
Journal of food composition and analysis, 18(8), 751-758 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service