Skip to Content
Merck
All Photos(2)

Key Documents

854999

Sigma-Aldrich

2′-Deoxyguanosine hydrate

99%

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$112.00
1 G
$326.00

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$112.00
1 G
$326.00

About This Item

Empirical Formula (Hill Notation):
C10H13N5O4 · xH2O
CAS Number:
Molecular Weight:
267.24 (anhydrous basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.22

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

powder

optical activity

[α]20/D −30°, c = 0.2 in H2O

mp

>300 °C (dec.) (lit.)

SMILES string

[H]O[H].NC1=Nc2c(ncn2[C@H]3C[C@H](O)[C@@H](CO)O3)C(=O)N1

InChI

1S/C10H13N5O4.H2O/c11-10-13-8-7(9(18)14-10)12-3-15(8)6-1-4(17)5(2-16)19-6;/h3-6,16-17H,1-2H2,(H3,11,13,14,18);1H2/t4-,5+,6+;/m0./s1

InChI key

LZSCQUCOIRGCEJ-FPKZOZHISA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Camila Donato Mahl et al.
Diagnostic microbiology and infectious disease, 82(3), 203-208 (2015-05-04)
In this study, we assessed the generation of reactive oxygen species (ROS) induced by subinhibitory concentration of fluconazole in susceptible and resistant Candida glabrata strains at stationary growth phase and measured their oxidative responses parameters: glutathione peroxidase (GPx), superoxide dismutase
Riccardo Vanzo et al.
Cell death and differentiation, 27(3), 1134-1153 (2019-08-15)
Autophagy is an evolutionarily conserved process that captures aberrant intracellular proteins and/or damaged organelles for delivery to lysosomes, with implications for cellular and organismal homeostasis, aging and diverse pathologies, including cancer. During cancer development, autophagy may play both tumour-supporting and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service