Skip to Content
Merck
All Photos(2)

Key Documents

852228

Sigma-Aldrich

2′-Deoxyguanosine 5′-monophosphate disodium salt hydrate

≥98%

Synonym(s):

2′-Deoxy-5′-guanylic acid disodium salt, 5′-dGMP disodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$253.00

$253.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$253.00

About This Item

Empirical Formula (Hill Notation):
C10H12N5Na2O7P · xH2O
CAS Number:
Molecular Weight:
391.18 (anhydrous basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.22

$253.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

Quality Level

Assay

≥98%

form

solid

impurities

≤4% ethanol

mp

>245 °C (dec.) (lit.)

SMILES string

[Na+].[Na+].[H]O[H].NC1=Nc2c(ncn2[C@H]3C[C@H](O)[C@@H](COP([O-])([O-])=O)O3)C(=O)N1

InChI

1S/C10H14N5O7P.2Na.H2O/c11-10-13-8-7(9(17)14-10)12-3-15(8)6-1-4(16)5(22-6)2-21-23(18,19)20;;;/h3-6,16H,1-2H2,(H2,18,19,20)(H3,11,13,14,17);;;1H2/q;2*+1;/p-2/t4-,5+,6+;;;/m0.../s1

InChI key

HQSJCEYJAGVPJG-BIHLCPNHSA-L

Application

Reactant involved in:
  • Analysis of self-assembling in solution and nucleation / growth of G-qudruplexes
  • Nucleophilic trapping
  • Reductive alkylation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingfeng Li et al.
PloS one, 8(1), e54420-e54420 (2013-02-01)
Free extracellular DNA provides nutrition to bacteria and promotes bacterial evolution by inducing excessive mutagenesis of the genome. To understand the influence of extracellular DNA fragments on D. radiodurans, we investigated cell growth and survival after extracellular DNA or dNMPs
Mariko Higuchi et al.
Journal of structural biology, 173(1), 20-28 (2010-10-05)
MutT distinguishes substrate 8-oxo-dGTP from dGTP and also 8-oxo-dGMP from dGMP despite small differences of chemical structures between them. In this paper we show by the method of molecular dynamics simulation that the transition between conformational substates of MutT is
Yancheng Liu et al.
Photochemistry and photobiology, 88(3), 639-644 (2012-02-11)
Laser flash photolysis studies have been carried out to investigate the reactions of ciprofloxacin (CPX) with 2'-deoxyguanosine-5'-monophosphate (dGMP), N, N, N', N'-tetramethyl-p-phenylenediamine (TMPD) and ferulic acid (FCA) in neutral aqueous solutions, respectively. CPX triplet state ((3)CPX*) can be quenched by
Jean L Whittingham et al.
The Biochemical journal, 428(3), 499-509 (2010-04-01)
Plasmodium falciparum is the causative agent of malaria, a disease where new drug targets are required due to increasing resistance to current anti-malarials. TMPK (thymidylate kinase) is a good candidate as it is essential for the synthesis of dTTP, a
Mahmoud Kandeel et al.
Nucleic acids symposium series (2004), (53)(53), 39-40 (2009-09-15)
In Plasmodium falciparum, deoxyguanylate was found to be a substrate for several DNA metabolizing enzymes. Guanylate kinase utilizes dGMP with very low specificity, which is estimated to be the lowest among well-known prokaryotic and eukaryotic enzymes. Furthermore, thymidylate kinase, which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service