Skip to Content
Merck
All Photos(1)

Key Documents

286931

Sigma-Aldrich

Pentacosane

99%

Synonym(s):

n-Pentacosane

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$164.00
5 G
$603.00

$164.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$164.00
5 G
$603.00

About This Item

Linear Formula:
CH3(CH2)23CH3
CAS Number:
Molecular Weight:
352.68
Beilstein:
1763638
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$164.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

bp

169-170 °C/0.05 mmHg (lit.)

mp

53-56 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C25H52/c1-3-5-7-9-11-13-15-17-19-21-23-25-24-22-20-18-16-14-12-10-8-6-4-2/h3-25H2,1-2H3

InChI key

YKNWIILGEFFOPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentacosane is one of the major components of the essential oils obtained from the leaves of Malus domestica tree[1], dried flower of Malva sylvestris L[2] and Aerva javanica (Burm.f.) Juss. ex Schult. Seed[3].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhammad Qasim Samejo et al.
Pakistan journal of pharmaceutical sciences, 26(4), 757-760 (2013-07-03)
Aerva javanica (Burm.f.) Juss. ex Schult. seed essential oils were obtained by hydrodistillation (HD) and dry steam distillation (SD) extracting methods and analyzed by using gas chromatography-mass spectrometry(GC-MS). Twenty and eighteen components representing 90.5% and 95.6% of the seed essential
Gianluca Gilardoni et al.
Plants (Basel, Switzerland), 9(10) (2020-10-25)
An essential oil was distilled with 0.25% yield from fresh flowers of Dalea mutisii Kunth, a native species mainly growing in the Andean region of Ecuador. A total of 50 compounds were identified, and most of them were quantified. The
Atsushi Usami et al.
Journal of oleo science, 62(8), 563-570 (2013-08-30)
In this study, the aroma-active compounds in the dried flower of Malva sylvestris L. were extracted by hydrodistillation and analyzed by gas chromatography-mass spectrometry (GC-MS), and gas chromatography-olfactometry (GC-O) and aroma extraction dilution analysis (AEDA). A light yellow oil with
Mayanka Walia et al.
Evidence-based complementary and alternative medicine : eCAM, 2012, 649727-649727 (2012-05-24)
Light pale-colored volatile oil was obtained from fresh leaves of Malus domestica tree, growing in Dhauladhar range of Himalaya (Himachal Pradesh, India), with characteristic eucalyptol dominant fragrance. The oil was found to be a complex mixture of mono-, sesqui-, di-terpenes
Megumi Iwasa et al.
Journal of oleo science, 64(8), 825-833 (2015-07-17)
The chemical composition of essential oil extracted from Uncaria Hook ("Chotoko" in Japanese), the branch with curved hook of the herbal medicine Uncaria rhynchophylla has been investigated by GC and GC-MS analyses. Eighty-four compounds, representing 90.8% of the total content

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service